Would you like to log in again?
3M™ Aqua-Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge AP-DW80/90
Supplier: 3M Healthcare
Aqua-Pure™
7100000550
76030-223CS 2002.75 USD InStock
76030-223
3M™ Aqua-Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge AP-DW80/90
Filters Filter Cartridges
3M™ Aqua–Pure™ under sink dedicated faucet replacement water filter cartridge (AP–DW80/90) is suitable for the AP–DWS1000 and AP–DWS1000 LF filtration systems. It is NSF certified to reduce particulate, chlorine taste and odor, lead, turbidity, parasitic protozoan cysts, VOCs and MTBE and deliver cleaner, clearer, better tasting water through a dedicated faucet (included in the AP–DWS1000).
  • Reduces particulate, chlorine taste and odor, lead, turbidity, parasitic protozoan cysts, VOCs and MTBE
  • Advanced dual stage carbon–block filtration process to help provide cleaner, clearer better tasting drinking water
  • 625 gallons/2366 L capacity
  • Sanitary Quick Change (SQC) cartridge design makes cartridge change–outs fast and easy
The 3M™ Aqua–Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge AP–DW80/90 provides water filtration on demand for drinking (water, coffee, tea, baby formula, etc.) and for cooking through our advanced dual stage carbon–block filtration process
Best Features: 0.6 gpm/2.27 lpm flow rate, 25 to 125 psi/172 to 862 kPa pressure range, 4.4 to 37.8 °C (40 to 100 °F).
In many filtration applications, carbon is often the preferred media of choice to absorb certain impurities and one of the most powerful absorbents available. Carbon is highly effective due to the vast surface area contained within a small footprint. One pound of carbon contains a surface area of approximately 100 to 125 acres and can absorb an extremely wide range of substances which may be in drinking water. To maximize system performance and efficiency, regularly scheduled maintenance including replacement of the filter cartridges at the required interval, at the rated capacity, or sooner if a reduction in water flow occurs is required. Filter life is dependent on actual use and incoming water quality.
The AP–DW80/90 replacement water filter cartridge set allows for use on both chlorinated (municipal) and non–chlorinated (well water) systems.
Certifications: NSF certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty