Would you like to log in again?
3M™ High Flow Series Replacement Cartridge, Model HF30, 5615105
Supplier: 3M Healthcare
3M™
7000001673
76030-329CS 1145.29 USD InStock
76030-329
3M™ High Flow Series Replacement Cartridge, Model HF30, 5615105
Filters Filter Cartridges
3M™ Water Filtration Products High Flow Series Model HF30 Replacement Cartridge for cold beverage applications and provide you with Recipe Quality Water™ for a consistent quality carbonated and non–carbonated beverage that can help increase repeat business and elevate profitability.
  • Provides consistent quality water for cold beverages by reducing sediment*, chlorine taste and odor and cyst**
  • Enhances water clarity for visually appealing carbonated and non–carbonated beverages
  • Reduction of sediment helps prevent equipment wear and can help extend the life of pumps, valves, o–rings and seals
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs
Our 3M™ Water Filtration Products High Flow Series Model HF30 Replacement Cartridge for cold beverage applications is designed to be effective at reducing sediment*, chlorine taste and odor and cyst** allowing you to feel more confident in the quality of the products you serve.
The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change–outs that are sanitary by design – requiring no contact with the filter media during cartridge change–out.
3M™ Water Filtration Products branded replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you're giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point–of–entry to dedicated point–of–use water filtration systems, look to 3M Purification to help you find the right solution.
Ordering information: **As tested with E. Coli ATCC (11229). Tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty