Would you like to log in again?
3M™ Aqua-Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP35R
Supplier: 3M Healthcare
Aqua-Pure™
7100049845
76030-589CS 2610.36 USD InStock
76030-589
3M™ Aqua-Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP35R
Filters Filter Cartridges
3M™ Aqua–Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP35R, which is compatible with the Everpure H–300 filter system as an activated carbon filter with a particulate reduction rating of 0.5 microns nominal. This cartridge design is suitable for drinking water, cooking water and food applications.
  • Advanced dual stage carbon–block filtration process to help provide cleaner, clearer better tasting drinking water
  • Reduces particulate, chlorine taste & odor, parasitic protozoan cysts, asbestos, lead, MTBE, Trihalomethanes (TTHM), VOCs (including benzene, atrazine lindane)
  • Compatible with the Everpure® H–300 drinking water system
  • Sanitary Quick Change (SQC) design allows for fast and easy cartridge change–outs without a filter wrench
Our 3M™ Aqua–Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP35R will also reduce chlorine taste and odor reduction at a flow rate of up to 0.5 gpm /1.89 lpm to provide cleaner, clear drinking water, cooking water for food applications.
Best features: Advanced dual stage carbon–block filtration, 350 gallons/1325 L capacity, Quick change cartridges are sanitary in design, requiring no contact with the filter media during cartridge change–out, Compatible with the Everpure H–300 drinking water system, 25 to 125 psi / 172 to 862 kPa operating pressure and 4.4 to 37.8 °C (40 to 100 °F) temperature range.
In many filtration applications, carbon is often the preferred media of choice to absorb certain impurities and one of the most powerful absorbents available. Carbon is highly effective due to the vast surface area contained within a small footprint. One pound of carbon contains a surface area of approximately 100 to 125 acres and can absorb an extremely wide range of substances which may be in drinking water.
It's specially formulated blend of high adsorption carbon media reduces unwanted chlorine taste and odor and reduces particulate down to the 0.5 µm (nominal) level. This cartridge, also helps reduce water–borne parasitic protozoan cysts. Its design is compatible with current and previous Everpure H–300 systems but requires fewer change–outs. Sanitary Quick Change (SQC) design, allows for fast and easy cartridge change–outs without having to contact the filter media or the need for a filter wrench. Less frequent cartridge change–outs as compared to typical drop–in style cartridges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty