Would you like to log in again?
3M™ High Flow Series Replacement Cartridge, Model HF60 Replacement Cartridge, 5613403
Supplier: 3M Healthcare
3M™
7000001666
76031-662EA 571.11 USD InStock
76031-662
3M™ High Flow Series Replacement Cartridge, Model HF60 Replacement Cartridge, 5613403
Filters Filter Cartridges
3M™ High Flow Series Replacement Cartridge, Model HF60 Replacement Cartridge provides consistent high quality water by particulate*, cyst**, bacteria1, and chlorine taste and odor.
  • Provides consistent high quality water
  • Reduces particulate*, cyst**, bacteria1, and chlorine taste and odor from incoming water
Our 3M™ High Flow Series Replacement Cartridge, Model HF60 Replacement Cartridge provides consistent high quality water by particulate*, cyst**, bacteria1, and chlorine taste and odor.
Excess particulate can cause abrasion and short life on pumps, seals, o–ings, and valves, while excess chlorine can cause unpleasant–tasting beverages. Integrated Membrane Pre–Activated Carbon Technology (“I.M.P.A.C.T.") filtration media combines a membrane with a premium activated carbon block to provide excellent throughput and cartridge life. The Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench.
Creative Filtration Technology 3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Recipe Quality Water™ Having cleaner and clearer water for your business is both a pleasure and a reassurance that you’re giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point–of–entry to dedicated point–of–use water filtration systems, look to 3M Purification to help you find the right solution.
Ordering information: **Based on the use of Cryptosporidium parvum oocysts.
1As tested with E. Coli ATCC (11229). Tested and verified by manufacturer’s laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty