Would you like to log in again?
3M™ Replacement Cartridge (Original CUNO 8000 Series), Model SW1350, 5634201
Supplier: 3M Healthcare
3M™
7100039086
76030-635CS 937.06 USD InStock
76030-635
3M™ Replacement Cartridge (Original CUNO 8000 Series), Model SW1350, 5634201
Filters Filter Cartridges
3M™ Water Filtration Products Model SWC1350 (Original CUNO 8000 Series) Scale Reduction Softening Replacement Cartridge works to soften hard water while helping to provide equipment protection against the potentially harmful effects of scale build-up. The Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs.
  • Reduces scale*, helping to provide an additional layer of equipment protection from the potentially harmful effects of scale build-up
  • Softening cartridge is specially designed for use with hard water
  • Specifically designed for point-of-use foodservice applications
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
Our 3M™ Water Filtration Products Model SWC1350 (Original CUNO 8000 Series) Scale Reduction Softening Replacement Cartridge is designed to help provide an additional layer of equipment protection from the potentially harmful effects of scale build-up.
The Sanitary Quick Change (SQC) encapsulated design allows for fast and easy cartridge change-outs without the need for a filter wrench.
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Recipe Quality Water™.
A History of Innovation
For more than a century, 3M has applied innovation and technology to improve our customers' lives and support their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. Across thousands of products, the 3M brand represents consistency, superior quality, and value. Our success begins with a culture of creative collaboration that inspires a never ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies - often in combination - to an endless array of real-world customer needs. Of course, all of this is made possible by the people of 3M and our singular commitment to make life easier and better for people around the world. Toward that end, 3M is committed to providing innovative, cost-effective products that help protect people, equipment, and our environment. 3M Purification Inc. (formerly CUNO Incorporated) provides filtration and separation solutions for residential, foodservice, science labs, industrial, marine, and other applications.
*Tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty