Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3M™ Aqua-Pure™ Whole House Standard Sump Replacement Water Filter Drop-in Cartridge AP124

Supplier: 3M Healthcare
Aqua-Pure™
5620601 7000051058 7000051059
76030-481CS 424.69 USD InStock
76030-481 76030-480 76030-485
3M™ Aqua-Pure™ Whole House Standard Sump Replacement Water Filter Drop-in Cartridge AP124
Filters Filter Cartridges
3M™ Aqua–Pure™ whole house standard diameter water filter drop–in replacement cartridge AP124 for the AP100 and standard and large diameter stainless steel housing series, is suitable for water filtration. This standard 2.5" diameter by 9.8" length drop–in cartridge is manufacturer tested and verified to reduce sediment down to 50 µm nominal.

  • Cartridge reduces sediment
  • Average cartridge life: 6 months (incoming water quality may affect cartridge life)
  • Manufactured using proprietary polypropylene manufacturing process
  • Grooved surface increases total surface area for longer cartridge life
  • Rigid construction helps to eliminate sediment 'unloading' due to variances in water pressure

Features include: Flow Rates: up to to 8 gpm/30.28 lpm (housing dependent), Average life: 6 months (incoming water quality may affect cartridge life), 50 µm nominal, 2.5"/6.35 cm by 9.8"/24.9 cm, 25 to 125 psi/172 to 862 kPa pressure range, 4.4...37.8 °C (40...100 °F) temperature range.

To maximize system performance and efficiency, regularly scheduled maintenance including replacement of the filter cartridges at the required interval, at the rated capacity, or sooner if a reduction in water flow occurs is required. Filter life is dependent on actual use and incoming water quality.

Certifications: Materials are FDA CFR–21 compliant.

Caution: Tested and verified by manufacturer's laboratory.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty