Would you like to log in again?
3M™ OCS Office Coffee Replacement Cartridge, Model CS-11, 5589501
Supplier: 3M Healthcare
3M™
7000050382
76030-231CS 903.96 USD InStock
76030-231
3M™ OCS Office Coffee Replacement Cartridge, Model CS-11, 5589501
Filters Filter Cartridges
3M™ Water Filtration Products Model CS-11 OCS Office Coffee Replacement Cartridge provides you with consistent quality water for hot beverages by reducing undesirable sediment* chlorine taste and odor and scale* from incoming water.
  • Provides consistent quality water for office coffee and hot cup applications by reducing sediment* and chlorine taste and odor
  • Reduces the ability of calcium and magnesium to form as hard scale* helping to reduce downtime and unscheduled service maintenance
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
Our 3M™ Water Filtration Products Model CS-11 OCS Office Coffee Replacement Cartridge is designed to be effective at reducing sediment,* scale* and chlorine taste and odor allowing you to feel more confident in the quality of the products you serve.
The Sanitary Quick Change (SQC) design allows for fast and easy cartridge change-outs without the need for a filter wrench and are sanitary by design, requiring no contact with the filter media during cartridge change-out.
3M™ Water Filtration Products replacement cartridges and filter systems offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust and quality around the world.
Recipe Quality Water™.
A History of Innovation
For more than a century, 3M has applied innovation and technology to improve our customers' lives and support their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. Across thousands of products, the 3M brand represents consistency, superior quality and value. Our success begins with a culture of creative collaboration that inspires a never ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies - often in combination - to an endless array of real-world customer needs. Of course, all of this is made possible by the people of 3M and our singular commitment to make life easier and better for people around the world. Toward that end, 3M is committed to providing innovative, cost-effective products that help protect people, equipment and our environment. 3M Purification Inc. (formerly CUNO Incorporated), provides filtration and separation solutions for residential, food service, science labs, industrial, marine and other applications.
*Tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty