Would you like to log in again?
3M™ Aqua-Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP15
Supplier: 3M Healthcare
Aqua-Pure™
7100090101
76030-585CS 1559.17 USD InStock
76030-585
3M™ Aqua-Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP15
Filters Filter Cartridges
The 3M™ Aqua–Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP15 is compatible with the Everpure H–100 Under Sink Water Filter System. This retrofit under sink replacement cartridge is NSF certified to reduce particulate, chlorine taste and odor and parasitic protozoan cysts.
  • Reduces particulate, chlorine taste/odor and parasitic protozoan cysts
  • Advanced dual stage carbon–block filtration process helps provide cleaner, clearer, better tasting drinking water
  • Sanitary Quick Change (SQC) cartridge design makes cartridge change–out fast and easy
  • Retrofit cartridge works with the Everpure® H–100 drinking water system
Our 3M™ Aqua–Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP15 is compatible with the Everpure H–100 uses advanced dual stage carbon–block filtration.
Best Features: 0.5 gpm/1.89 lpm flow rate, 2000 gallon/7571 L capacity, 25 to 125 psi/172 to 862 kPa pressure range, 4.4 to 37.8 °C (40 to 100 °F) temperature range. To maximize system performance and efficiency, regularly scheduled maintenance including replacement of the filter cartridges at the required interval, at the rated capacity, or sooner if a reduction in water flow occurs is required. Filter life is dependent on actual use and incoming water quality.
Best features: Sanitary Quick Change (SQC) design allows to easily change out and replace the encapsulated media cartridges with a simple quarter turn without touching the spent filter media or contaminated water. Aqua–Pure™ EP15 Under Sink Dedicated Faucet Replacement Water Filter Cartridges, working together with other parts, is certified to reduce many contaminants which may be in drinking water.
Certifications: The retrofit under sink replacement cartridge is NSF Certified to reduce particulate, chlorine taste and odor and parasitic protozoan cysts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty