Would you like to log in again?
3M™ High Flow Series Replacement Cartridge, Model HF60-CL, 5625901
Supplier: 3M Healthcare
3M™
7100045318
76031-700EA 616.13 USD InStock
76031-700
3M™ High Flow Series Replacement Cartridge, Model HF60-CL, 5625901
Filters Filter Cartridges
3M™ High Flow Series Replacement Cartridge, Model HF60–CL provides consistent high quality water by reducing particulate*, chlorine taste and odor, chloramines, cyst**, and bacteria. The Sanitary Quick Change (SQC) design allows for easy cartridge change–outs.
  • Provides consistent high quality water
  • Reduces particulate*, chlorine taste and odor, chloramines, cyst**, and bacteria from incoming water
  • Built–in scale inhibition can help to prevent the potentially harmful effects of scale build–up on equipment
Our 3M™ High Flow Series Replacement Cartridge, Model HF60–CL provides consistent quality water by reducing particulate*, chlorine taste and odor, chloramines, cyst**, and bacteria.
Excess particulate can cause abrasion and short life on pumps, seals, o–rings, and valves, while excess chlorine and chloramines residual can cause unpleasant tasting beverages. Integrated Membrane Pre–Activated Carbon Technology (“I.M.P.A.C.T.") filtration media combines a membrane with a premium activated carbon block to help provide excellent throughput and cartridge life. Our Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench.
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you’re giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point–of–entry to dedicated point–of–use water filtration systems, look to 3M Purification to help you find the right solution.
Ordering information: **Based on the use of cryptosporidium parvum oocysts.
1As tested with E. Coli ATCC (11229). Tested and verified by manufacturer’s laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty