Would you like to log in again?
3M™ High Flow Series Replacement Cartridge, Model HF60-CLS, 5625902
Supplier: 3M Healthcare
3M™
7000125910
76030-500EA 640.83 USD InStock
76030-500
3M™ High Flow Series Replacement Cartridge, Model HF60-CLS, 5625902
Filters Filter Cartridges
3M™ High Flow Series Replacement Cartridge, Model HF60–CLS provides consistent high quality water by reducing particulate*, chlorine taste and odor, chloramines, scale*, cyst**, and bacteria. The Sanitary Quick Change (SQC) design allows for easy cartridge change–outs.
  • Provides consistent high quality water
  • Reduces particulate*, chlorine taste and odor, chloramines, cyst**, and bacteria1 from incoming water
  • Built–in scale inhibition can help to prevent the potentially harmful effects of scale build–up on equipment
Our 3M™ High Flow Series Replacement Cartridge, Model HF60–CLS provides consistent quality water by reducing particulate*, chlorine taste and odor, chloramines, cyst**, and bacteria1.
Excess particulate can cause abrasion and short life on pumps, seals, o–rings, and valves, while excess chlorine and chloramines residual can cause unpleasant tasting beverages. Built–in scale inhibition can help to prevent the potentially harmful effects of scale build–up on equipment. Integrated Membrane Pre–Activated Carbon Technology ('I.M.P.A.C.T.') filtration media combines a membrane with a premium activated carbon block to help provide excellent throughput and cartridge life. Our Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench.
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you’re giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point–of–entry to dedicated point–of–use water filtration systems, look to 3M Purification to help you find the right solution.
For more than a century, 3M has applied innovation and technology to improve our customers’ lives and support their business goals. From our headquarters in St. Paul, Minnesota, we serve industrial and commercial customers and consumers in nearly 200 countries. Across thousands of products, the 3M brand represents consistency, superior quality, and value. Our success begins with a culture of creative collaboration that inspires a never–ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies – often in combination – to an endless array of real–world customer needs. Of course, all of this is made possible by the people of 3M and our singular commitment to make life easier and better for people around the world. Toward that end, 3M is committed to providing innovative, cost–effective products that help protect people, equipment and our environment. 3M Purification Inc. (formerly CUNO Incorporated) provides filtration and separation solutions for residential, foodservice, science labs, industrial, marine and other applications.
1As tested with E. Coli ATCC (11229). Tested and verified by manufacturer’s laboratory.
Ordering information: **Based on the use of cryptosporidium parvum oocysts.
Caution: Based on the use of cryptosporidium parvum oocysts. ** As tested with E. Coli ATCC (11229). Tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty