Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
3M™ Aqua-Pure™ Whole House Sanitary Quick Change Replacement Water Filter Cartridge AP917HD
Supplier: 3M Healthcare
Aqua-Pure™
7100039299
76028-692EA 715.53 USD InStock
76028-692
3M™ Aqua-Pure™ Whole House Sanitary Quick Change Replacement Water Filter Cartridge AP917HD
Filters Filter Cartridges
3M™ Aqua–Pure™ Whole House Sanitary Quick–Change Replacement Water Filter Cartridge AP917HD for the AP903 system as an activated carbon filter with a nominal heavy duty particulate reduction rating of 5 µm and a 100000 gallon capacity. This cartridge design is suitable for use on both chlorinated (municipal) and non–chlorinated (well water) systems.
  • Replacement cartridge provides up to 100000–gallon capacity, approximately one–year cartridge life, dependent upon water quality conditions
  • Up to 10 gallons per minute (GPM) flow rate
  • Sanitary Quick Change (SQC) design allows for fast and easy cartridge change–outs without a filter wrench
Our 3M™ Aqua–Pure™ Whole House Sanitary Quick–Change Replacement Water Filter Cartridge AP917HD for the AP903 system reduces sediment, chlorine taste and odor at a flow rate of up to 10 GPM to meet the needs of large homes with at least 2.5 bathrooms.
Features include: Filter cartridge is O–ring seal type, Maximum flow rate is up to 20 gpm/75.7 lpm*, AP917HD Filter Cartridge incorporates carbon block filtration with a non–woven pleat media, Cartridges are sanitary in design, requiring no contact with the filter media during cartridge change–out.
3M, a household brand name, is a known leader in filtration technology and recognized for innovation, trust and quality around the world and 3M branded water filtration products have been addressing water quality issues for over 50 years. 3M products offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are a quality advantage we strive to provide on many 3M water quality improvement solutions.
Having clean and clear drinking water in your home is as pleasurable as it is reassuring to know that you're giving your family the best. With a water filtration system from 3M Purification, you can provide filtered drinking water for every member of your family. From whole home water filtration to drinking water systems, look to 3M Purification to help you find the right solution.
For more than a century, 3M has applied innovation and technology to improving our customers’ lives and supporting their business goals. Today the company, from its headquarters in St. Paul, Minnesota, serves industrial and commercial customers and consumers in nearly 200 countries. The 3M brand, across thousands of products, represents consistency, superior quality, and value. Our success begins with our culture of creative collaboration that inspires a never–ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies – often in combination – to an endless array of real–world customer needs. Of course, all of this made possible by the people of 3M, and their singular commitment to make life easier and better for people around the world. 3M is committed to providing innovative, cost–effective products that help protect people, equipment, and our environment. 3M Purification Inc. (formerly CUNO Incorporated), provides filtration and separation solutions for residential, food service, science labs, industrial, marine, and other applications.
Our innovative Sanitary Quick Change (SQC) design, allows for fast and easy cartridge change–outs without having to contact the filter media or the need for a filter wrench. Our Sanitary Quick Change (SQC) is one of the industry's most advanced whole house water filtration system features for this application and minimizes contact with the filter media.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty