Would you like to log in again?
3M™ Under Sink Reverse Osmosis Replacement Water Filter Cartridge 3MROP316
Supplier: 3M Healthcare
Aqua-Pure™
7100049298
76028-369CS 2837.51 USD InStock
76028-369
3M™ Under Sink Reverse Osmosis Replacement Water Filter Cartridge 3MROP316
Filters Filter Cartridges
3M™ 3MROP316 Under Sink Reverse Osmosis Replacement Water Filter Cartridge serves as the post-filter for reverse osmosis system. Cartridge is designed to reduce particulate, chlorine taste and odor, and lead as the final filtration stage and then deliver cleaner, clearer, better tasting water. Change-out is fast and easy with our Sanitary Quick Change (SQC) system.
  • Post-filter cartridge reduces particulate, and chlorine taste and odor
  • Sanitary Quick Change (SQC) cartridge design helps provide fast and easy cartridge change-out
  • Slim profile and space saving design reduces overall under cabinet footprint
  • Advanced multi-stage reverse osmosis water filtration process helps provide cleaner, better tasting drinking water through a dedicated faucet
Just Add Water.
Our 3M™ 3MROP316 Under Sink Reverse Osmosis Replacement Water Filter Cartridge is easily changed do to the Sanitary Quick Change (SQC) design. With our SQC encapsulated media cartridges, we eliminate contact with the filter media during cartridge change-outs with no need for a filter wrench. A quick turn of the filter is all that is needed to remove the old and replace with the new. The SQC design is one of the industry's most advanced water filtration system features. We also incorporated a built-in shut-off valve in the system that allows for changing the filter without shutting off the water supply.
Our 3MRO301 system installs under the kitchen sink and delivers cleaner, clearer, better tasting drinking water through a dedicated drinking water faucet. This system provides water filtration on demand for drinking water, coffee, tea, baby formula, etc. and cooking. Our universal system design allows for use on both chlorinated (municipal) and non-chlorinated (well water) systems.
Best Features.
Our reverse osmosis water filtration systems help to reduce impurities by forcing water through a membrane and leaving contaminants behind. Reverse osmosis is typically used in connection with thin film composite (TFC) membranes and activated carbon pre-filters and post-filters.
To maximize system performance and efficiency, regularly scheduled maintenance including replacement of the filter cartridges at the required interval, at the rated capacity, or sooner if a reduction in water flow occurs. Filter life is dependent on actual use and incoming water quality.
About Carbon Filtration
In many filtration applications, carbon is often the preferred media of choice to absorb certain impurities and one of the most powerful absorbents available. Carbon is highly effective due to the vast surface area contained within a small footprint. One pound of carbon contains a surface area of approximately 100 to 125 acres and can absorb an extremely wide range of substances which may be in drinking water.
Ideas Flow Like… Water
3M, a household brand name, is a known leader in filtration technology and recognized for innovation, trust and quality around the world and 3M branded water filtration products have been addressing water quality issues for over 50 years. 3M products offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are a quality advantage we strive to provide on many 3M water quality improvement solutions.
Water: Nothing’s Closer to Home
Having clean and clear drinking water in your home is as pleasurable as it is reassuring to know that you're giving your family the best. With a water filtration system from 3M Purification, you can provide filtered drinking water for every member of your family. From whole home water filtration to drinking water systems, look to 3M Purification to help you find the right solution.
A History of Innovation
For more than a century, 3M has applied innovation and technology to improving our customers’ lives and supporting their business goals. Today the company, from its headquarters in St. Paul, Minnesota, serves industrial and commercial customers and consumers in nearly 200 countries. The 3M brand, across thousands of products, represents consistency, superior quality, and value. Our success begins with our culture of creative collaboration that inspires a never ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies – often in combination – to an endless array of real-world customer needs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty