Would you like to log in again?
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9112EL, 5631701
Supplier: 3M Healthcare
3M™
7100049907
76030-595CS 1233.87 USD InStock
76030-595
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9112EL, 5631701
Filters Filter Cartridges
3M™ Water Filtration Products Model CFS9112EL Retrofit 9000 Series Replacement Cartridge provides Recipe Quality Water™ for consistent beverages by reducing undesirable sediment* and chlorine taste and odor. Extended length replacement cartridges fit most Everpure® commercial foodservice heads allowing you to choose from a variety of 3M™ Water Filtration Products commercial 9000 series cartridges.
  • Provides consistent quality water for commercial post mix machines by reducing sediment* and chlorine taste and odor
  • Enhances water clarity for visually appealing cold beverages
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
  • Extended Length retrofit cartridges provide higher gallon capacity and flow rates for increased production over the standard length 3M™ Water Filtration Products cartridges
Our 3M™ Water Filtration Products Model CFS9112EL Retrofit 9000 Series Replacement Cartridge is designed to be effective at reducing sediment* and chlorine taste and odor allowing you to feel more confident in the quality of the products you serve
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty