Would you like to log in again?
3M™ Aqua-Pure™ Under Sink Full Flow Water Filter Replacement Cartridge AP517
Supplier: 3M Healthcare
Aqua-Pure™
7100049866 AP51711
76031-263CS 3232.55 USD InStock
76031-263 76031-262
3M™ Aqua-Pure™ Under Sink Full Flow Water Filter Replacement Cartridge AP517
Filters Filter Cartridges
3M™ Aqua–Pure™ Under Sink Full Flow Water Filter Replacement Cartridge AP517 for the AP–510 filter system is suitable for water filtration. This under sink replacement cartridge reduces sediment and chlorine taste and odor and scale. This replacement cartridge is designed for use with the AP–510 Under Sink Full Flow Water Filter System.
  • Dual stage carbon–block filtration process helps provide cleaner, clearer better tasting drinking water
  • Sanitary Quick Change (SQC) cartridge design helps makes cartridge change–out fast and easy
  • Filter allows for use on both chlorinated (municipal) and non–chlorinated (well water) systems
Our 3M™ Aqua–Pure™ Under Sink Full Flow Water Filter Replacement Cartridge AP517 installs in the AP–510 Under Sink Full Flow Water Filter System and is good choice for delivering cleaner, clearer, better tasting drinking water
The dual stage carbon–block filtration process from this cartridge provides water filtration on demand for drinking (water, coffee, tea, baby formula, etc.) and for cooking purposes. This under sink full flow replacement water filter cartridge is manufacturer tested and verified to reduce sediment1 and chlorine taste and odor1 and scale1. The AP–510 system design, using the AP–517 replacement water filter cartridge allows for use on both chlorinated (municipal) and non–chlorinated (well water) systems. Best Features: 1.75 gpm/6.6 lpm flow rate, 2000 gallon/7571 L capacity. 25 to 125 psi/172 to 862 kPa pressure range,4.4 to 37.8 °C (40 to 100 °F)
To maximize system performance and efficiency, regularly scheduled maintenance including replacement of the filter cartridges at the required interval, at the rated capacity, or sooner if a reduction in water flow occurs is required. Filter life is dependent on actual use and incoming water quality.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty