Would you like to log in again?
3M™ ScaleGard™ HP Pre-Filter Replacement Cartridge, Model HF90-CL-RO, 5613529
Supplier: 3M Healthcare
ScaleGard™
7100081270
76028-662EA 904.78 USD InStock
76028-662
3M™ ScaleGard™ HP Pre-Filter Replacement Cartridge, Model HF90-CL-RO, 5613529
Filters Filter Cartridges
3M™ ScaleGard™ HP Pre-Filter Replacement Cartridge, Model HF90-CL-RO provides consistent high quality water by reducing particulate, chlorine taste and odor, chloramines, cyst*, and bacteria. The Sanitary Quick Change (SQC) design allows for easy cartridge change-outs without the need for a filter wrench. This product is designed for use with 3M™ ScaleGard™ HP reverse osmosis systems.
  • Provides consistent high quality-water
  • Reduces particulate, chlorine taste and odor, chloramines, cyst*, and bacteria1 from incoming water
  • Integrated Membrane Pre-Activated Carbon Technology (“I.M.P.A.C.T.”) filtration media combines a membrane with a premium activated carbon block to help provide excellent throughput and cartridge life
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench
  • Replacement cartridge for 3M™ ScaleGard™ HP Reverse Osmosis Systems
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty