Would you like to log in again?
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9112-S, 5631604
Supplier: 3M Healthcare
3M™
7100045148
76030-577CS 3162.95 USD InStock
76030-577
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9112-S, 5631604
Filters Filter Cartridges
3M™ Water Filtration Products Model CFS9112-S Retrofit 9000 Series Replacement Cartridge provides Recipe Quality Water™ for clear and consistent quality ice, coffee and tea by reducing undesirable sediment*, chlorine taste and odor, and scale*. This cartridge fits most Everpure® commercial foodservice heads, allowing you to choose the 9000 Series cartridge that best suits your filtration needs.
  • Provides consistent quality water for commercial ice machines and coffee and tea brewers by reducing sediment*, chlorine taste and odor and scale*
  • Enhances water clarity for visually appealing and better tasting ice
  • Reduces the ability of calcium and magnesium to form as hard scale* on evaporator plates and heating elements, which can reduce downtime and unscheduled service maintenance
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
Our 3M™ Water Filtration Products Model CFS9112-S Retrofit 9000 Series Replacement Cartridge is designed to be effective at reducing sediment*, chlorine taste and odor, and scale* allowing you to feel more confident in the quality of the products you serve
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty