Would you like to log in again?
3M™ High Flow Series Replacement Cartridge, Model HF65, 5613407
Supplier: 3M Healthcare
3M™
7100006231
76031-666EA 544.11 USD InStock
76031-666
3M™ High Flow Series Replacement Cartridge, Model HF65, 5613407
Filters Filter Cartridges
3M™ High Flow Series Replacement Cartridge, Model HF65 provides consistent quality water by reducing particulate* and chlorine taste and odor.
  • Provides consistent quality water
  • Reduces particulate* and chlorine taste and odor from incoming water
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench
Our 3M™ High Flow Series Replacement Cartridge, Model HF65 provides consistent quality water by reducing particulate* and chlorine taste and odor
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty