Would you like to log in again?
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9005-IMF for Ice Maker Applications, 5593707
Supplier: 3M Healthcare
3M™
7100042484
76030-241CS 1492.86 USD InStock
76030-241
3M™ Replacement Cartridge Retrofit 9000 Series, Model CFS9005-IMF for Ice Maker Applications, 5593707
Filters Filter Cartridges
3M™ Water Filtration Products Model CFS9005-IMF Retrofit 9000 Series Ice Maker Filter Replacement Cartridge provides an additional layer of equipment protection from the potentially harmful effects of scale build-up. The Sanitary Quick Change (SQC) encapsulated design allows for fast and easy cartridge change-outs. This filter cartridge is compatible with most Everpure® foodservice heads.
  • Scale inhibitor provides an extra layer of equipment protection from the potentially harmful effects of scale build-up
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs
  • Various 3M™ Water Filtration Products 9000 Series retrofit cartridges are available to meet particular water quality concerns
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty