Would you like to log in again?
3M™ Aqua-Pure™ Whole House Sanitary Quick Change Replacement Water Filter Cartridge AP910R
Supplier: 3M Healthcare
Aqua-Pure™
7100007625
76028-690EA 568.98 USD InStock
76028-690
3M™ Aqua-Pure™ Whole House Sanitary Quick Change Replacement Water Filter Cartridge AP910R
Filters Filter Cartridges
3M™ Aqua–Pure™ whole house sanitary quick change replacement water filter cartridge AP910R is the replacement cartridge for the AP902 system. The universal design allows for use on both chlorinated (municipal) and non–chlorinated (well water) systems.
  • AP910R Activated carbon filters reduce particulate down to 15 µm nominal for all incoming water
  • Sanitary Quick Change (SQC) design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • Up to 10 gallons per minute (GPM) flow rate
  • Whole house water filtration helps reduce sediment build–up in water heaters and appliances
The 3M™ Aqua–Pure™ whole house sanitary quick change water filter system AP902 is a high performing, filter that reduces sediment.
This system reduces sediment for improved water quality while supplying cleaner, clearer water to every water source point in the house. Maintenance of the AP902 whole–house water filter system is as simple as replacing a light bulb. Our Sanitary Quick Change (SQC) design allows you to perform fast and easy cartridge change–outs, without the need for a filter wrench, with a simple quarter turn of the cartridge.
Features include: Maximum flow rates up to 20 gpm/75.7 lpm*, 25 to 125 psi/172 - 862 kPa pressure range, 37.8 °C (40 to 100 °F) temperature range, Non–woven pleat media surrounding activated carbon block reduces particulates down to 15 microns nominal at 10 gallons per minute.
Ordering information: *As tested by manufacturer’s laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty