Would you like to log in again?
3M™ Aqua-Pure™ Whole House Standard Sump Replacement Water Filter Drop-in Cartridge AP420
Supplier: 3M Healthcare
Aqua-Pure™
7100040509
76030-083CS 2193.54 USD InStock
76030-083
3M™ Aqua-Pure™ Whole House Standard Sump Replacement Water Filter Drop-in Cartridge AP420
Filters Filter Cartridges
3M™ Aqua–Pure™ whole house standard diameter water filter drop–in replacement cartridge AP420 for the AP100 and standard and large diameter stainless steel Housing Series, is a good choice in water filtration. This standard 2.75" diameter by 9.8" length drop–in cartridge is manufacturer tested and verified to reduce sediment down to 5 µm nominal.
  • Reduces sediment and scale
  • Average cartridge life: 6 months (incoming water quality may affect cartridge life)
  • Proprietary polypropylene manufacturing process
  • Grooved surface increases total surface area for longer cartridge life compared than non–grooved filter cartridges
  • Rigid construction helps to eliminate sediment 'unloading' due to variances in water pressure
The scale inhibition media helps prevent scale build–up on water heaters (both standard and tankless styles), as well as down–stream plumbing fixtures.
Additional features and specifications include: 5–micron nominal, Flow Rate: 10 gpm/37.9 gpm (housing dependent), 2.75"/7 cm by 9.8"/24.9 cm, 25 to 125 psi/172 to 862 kPa pressure range, 4.4 to 37.8 °C (40 to 100 °F) temperature range.
To maximize system performance and efficiency, regularly scheduled maintenance including replacement of the filter cartridges at the required interval, at the rated capacity, or sooner if a reduction in water flow occurs is required. Filter life is dependent on actual use and incoming water quality.
Ordering information: *Surfaces that are in contact with water are FDA CFR–21 compliant.
As tested and verified by manufacturer’s laboratory
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty