Would you like to log in again?
3M™ Water Factory Systems™ Under Sink Replacement Filter Cartridge
Supplier: 3M Healthcare
Water Factory Systems™
7100000557
76028-361CS 2914.48 USD InStock
76028-361
3M™ Water Factory Systems™ Under Sink Replacement Filter Cartridge
Filters Filter Cartridges
The 3M™ Water Factory Systems™ Under Sink Replacement Filter Cartridge FM 1500 reduces lead, cyst, sediment, chlorine odor and taste and delivers cleaner, better tasting water as part of an easy-to-use home filtration system.
  • Reduces lead, cyst, sediment, chlorine odor and taste
  • Compatible with corresponding Water Factory Systems Under Sink system for convenient use
  • Third party tested and approved for quality assurance
  • High capacity activated carbon block for higher absorption
  • Radial flow design offers the benefits of granular activated carbon (GAC) filtration, such as low pressure drop, while at the same time significantly reducing the release of carbon commonly associated with granular activated carbon style cartridges
  • Sanitary Quick Change (SQC) design eliminates need to handle the media
  • Provides high flow stream of drinking water through unique 'radial flow' cartridge design
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty