Would you like to log in again?
3M™ Zeta Plus™ U and UW Series Filter Cartridge
Supplier: 3M Healthcare
3M™ Zeta Plus™
7100085135 7010381335 7010413518 7010381741 7000125505 7010415234 7010409850 7000125506 7010381107 7000125623 7010413018 7010409023 7000144815 7000125622 7000050713 7010381161 7010411087 7010381110 7010413039 7100114488 7010410986
76034-527CS 1122.09 USD InStock
76034-527 76033-618 76034-515 76034-525 76034-513 76030-034 76034-520 76033-577 76033-565 76033-557 76033-579 76033-622 76033-569 76034-519 76033-616 76034-517 76034-528 76033-573 76033-575 76033-563 76028-318
3M™ Zeta Plus™ U and UW Series Filter Cartridge
Filters Filter Cartridges
Zeta Plus™ U and UW Series filters for the power generation industry are offered in two formulations. The U formulation is recommended for use with oils contaminated mainly with carbon particles, and the UW formulation is specifically formulated for use with oils contaminated with both carbon particles and moisture.
  • Products developed specifically for the filtration of Insulation oil used in power generation
  • Tight 10U and 10UW grades provide reduction of very fine particles
  • Water absorbent formulation provides superior water reduction capabilities (UW Grade)
Zeta Plus charge-modified filter media uses electrokinetic adsorption to remove contaminants and colloid materials smaller than those filtered by mechanical sieving alone.
Zeta Plus™ U and UW Series filter cartridges are used for reconditioning insulating oils in power generation. Two grades of media, 05 and 10, are available: 05 grade provides efficient reduction of particle contaminants in most insulation oil applications, while the 10 grade provides efficient reduction of ultrafine particles.
As oil specifications get tighter, finer filtration is required to meet those demands. The 10U formulation was developed to help reduce smaller contaminants such as fine carbon particulate.
Water contaminates the dielectric oil either from the atmosphere or breakdown of the oil itself. Water contamination is usually accompanied by particulate contamination, which then increases both the filtration and water reduction workload. Zeta Plus U and UW series formulations help address both problems.
The construction of Zeta Plus™ Series 05UW and 10UW cartridges allow it to plug on saturation with water, effectively shutting off flow. This feature is important for two reasons. First, it provides a clearly defined end-point to filter life. Second, the automatic shut-off helps prevent recontamination of the filtrate with water.
User-Friendly Design
Optional cartridge reduction baskets are available for manually or mechanically installing and removing cartridges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty