Would you like to log in again?
3M™ LifeASSURE™ SP Series Filter Cartridge
Supplier: 3M Healthcare
LifeASSURE™
7002501B020SP 7000125650
76029-141CS 4670.58 USD InStock
76029-141 76029-145
3M™ LifeASSURE™ SP Series Filter Cartridge
Filters Filter Cartridges
LifeASSURE™ 020SP series sterilizing grade filters and capsules are validated for absolute bacteria retention and provide relable sterile filtration performance.
  • Integrity test correlated with sterile filtration performance data and supported by Validation Guide
  • Robust cartridge construction, Cartridges withstand mechanical and thermal stress including Steam in Place operations
  • Cartridges 100% integrity tested
  • Ensures consistent performance to specifications, integrity testable pre- and post-use
  • Quality Systems Standard
  • Tested and optimized for pharmaceutical and biological processing
  • Materials of construction tested for biological safety (USP Class VI) and listed in Drug Master File (DMF) at FDA
The LifeASSURE 020SP series microporous membrane is charge modified to provide enhanced reduction of negatively charged biological contaminants such as endotoxin, virus and nucleic acid fragments. The combination of a validated bacteria rententive membrane, together with enhanced reduction of negatively charged contaminants, make the LifeASSURE 020SP series microporous membrane an ideal choice for pharmaceutical and biopharmaceutical sterilizing applications.
High area, pleated membrane design, High flow rates, low pressure drop, Smaller filter systems and longer service life for reduced filtration costs. Reliable 0.2 µm validated performance, Meets FDA definition of 0.2 µm sterilizing grade filter (107 CFU/cm² B. diminuta retention).
Charge-modified Nylon 6,6 filter medium, Retention of negatively-charged biological and particulate contaminants including endotoxins, Inherently hydrophilic membrane for easy wet-out. Low extractable levels, no surfactants or adhesives used in manufacturing, minimal effect on filtrate quality and purity.
Certifications: ISO 9001:2008 - registered quality system, quality management system approved by an accrediting body to an ISO 9001:2008
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty