Would you like to log in again?
3M™ Betafine™ PEG Series Filter Cartridge
Supplier: 3M Healthcare
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Betafine™
7010380397 7000001711 7010413430 7010338509 7000001835 7000050682 7000001724 7100111882 7010415616 7000050684 7010379646
76034-035CS 7392.36 USD InStock
76034-035 76034-033 76034-043 76034-021 76034-031 76034-041 76034-051 76034-029 76028-492 76034-027 76034-047
3M™ Betafine™ PEG Series Filter Cartridge
Filters Filter Cartridges
Betafine™ PEG series filter cartridge provides solutions for a variety of applications. This absolute–rated, graded–porosity 100% polypropylene pleated filter features our Advanced Pleat Technology (APT) that increases usable surface area and service life.
  • Advanced Pleat Technology Construction provides high surface area and higher product throughputs for prolonged service life
  • Absolute–rated filter performance for higher product quality and yields
  • Graded–porosity multi–layer filter media for higher contaminant holding capacity
  • Polypropylene cartridge components are free of adhesives and surfactants
  • Broad chemical compatibility for most aggressive process applications
Betafine PEG series filter cartridges effectively reduce the contaminating particles while maintaining or increasing process yields.
Advanced Pleat Technology (APT) design to increase the service life of every Betafine™ PEG series filter cartridge. This design technology maximizes the useful surface area while maintaining open flow paths between media pleats. The design also supports a graded–porosity media structure, which results in lower pressure drops, longer service life and lower overall filtration costs. These filters are manufactured using our staggered pleat configuration that, when combined with a novel support material, provides more open space between the pleats. The increased open area of the staggered pleats allows for greater contaminant loading between each pleat at the inside diameter, while reduced–length pleats take advantage of existing open space closer to the outside diameter. The result is a fully usable surface area that provides superior service life.
Applications: Electronics Manufacturing, Pre–reverse osmosis water filtration, Plating and cleaning baths, Complex multi–layer board processes, Fine Chemical Production, Clarification of high purity chemicals, Organic and inorganic chemical intermediates, Various acids and bases, Solvents and polymer solutions.
The Betafine™ PEG series filter cartridge's graded–porosity media structure reduces particles sequentially by size — the larger particles by the more open, outer medium and the smaller particles by the tighter, inner medium. The outer medium acts as a pre–filter, while the inner provides the absolute removal specified by the cartridge rating. Our unique construction effectively spreads the contaminant through the depth of the filter media resulting in high contaminant capacity with lower pressure drop for longer service life as compared to competitive filters. Fine chemical production demands economical, rigorous filter cartridges with long service life. Our durable polypropylene construction provides broad chemical and mechanical compatibility for many demanding process fluid applications. Compatibility is influenced by process operating conditions; in critical applications, cartridges should be tested under actual conditions to determine the correct selection.
These filter cartridges represent a major advance in pleated filter technology and performance that is built upon our history of filter design innovation. This absolute–rated, graded–porosity 100% polypropylene pleated cartridge features our highly effective Advanced Pleat Technology (APT) design that increases the usable filtration surface area while maintaining standard cartridge sizes. The result is a filter cartridge that has consistent performance and long service life.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty