Would you like to log in again?
3M™ Betafine™ PBG Series Filter Cartridge
Supplier: 3M Healthcare
Betafine™
7010415406 7010380873 7100129126 7010339241 7010338752 7010339881
76033-983CS 2267.01 USD InStock
76033-983 76033-985 76033-977 76033-987 76033-979 76033-981
3M™ Betafine™ PBG Series Filter Cartridge
Filters Filter Cartridges
3M™ Betafine™ PBG series filter cartridge is an absolute–rated, graded–porosity 100% polypropylene pleated filter cartridge that features advanced pleat technology (APT). APT increases the usable filtration service area while maintaining standard industrial cartridge dimensions.
  • Absolute rated filter performance
  • Graded–porosity multi–layer filter media captures contaminant throughout the filter media to help maximize filter life
  • Broad compatibility with solvents and cleaning solutions
It has a variety of applications – cleaning fluids, solvent streams, food fermentation, air and gas pre–filtration, as well as final filtration processes.
Applications: Pre–filtration before final membrane filtration, Final product clarification, Food fermentation feeds, intermediates and fermentation clarification, Blending water filtration, Cleaning fluids, Solvent streams, Air and gas pre–filtration and final filtration.
Our filter cartridge is constructed of FDA CFR–21 listed material. Betafine PBG series filter cartridges are available in a range of absolute micron ratings, from 0.2 to 10 µm. Proper use of our Betafine PBG Series filter cartridges reduce undesirable turbidity and haziness that could adversely affect the color, taste and overall quality of beverages.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty