Would you like to log in again?
3M™ 1M Series Filter Housing Part
Supplier: 3M Healthcare
CUNO™ 3M™
7000001590 7010379276 7000029178
76028-320CS 3843.25 USD InStock
76028-320 76028-324 76028-322
3M™ 1M Series Filter Housing Part
Filters Filter Cartridges
3M™ 1M Series Filter Housing parts are manufactured from high impact plastic.
  • DOE connection
  • 19.05 mm (¾") NPTF connections
  • Maximum operating pressure of 125 psi
Available with one–filter high and two–filter high transparent and opaque sumps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty