Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3M™ BHN Series Filter Housing

Supplier: 3M Healthcare

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

3M™
7000125821 7010291600 7100133130 7000125820 7000125819 7000125823
76031-736CS 1758.62 USD InStock
76031-736 76031-738 76031-732 76031-734 76031-740 76028-834
3M™ BHN Series Filter Housing
Filters Filter Cartridges
These economical BHN Series bag filter housings for applications not requiring ASME code housings. BHN Series housings, for use with #1 and #2 Size conventional bag filters or 3M™ DF series filter elements, are available in single–round configurations. These housings are either 304 or 316L stainless steel construction, including support basket and hold–down device for proper seating of filter.

  • Cost–effective designs help reduce capital investment costs
  • Compression sealing of filter bag keeps bag filters properly sealed in baskets
  • V–band clamp design with integral cover handle permits easy vessel opening and closing
  • Drains positioned at lowest point in housing to guard against residual build–up or fluid loss
  • Housing provides flexibility for using either a standard or specialty bag filter (3M™ DF Series Filter Elements)

3M™ BHN Series Housings are rated for operation up to 150 psi @ 250 °F.

3M Purification carries a complete line of filter housings for use in industrial applications offering quality, ease of use, flexibility, and economy.

Applications: Process waters, parts washers, machine coolants, paints and coatings, production wells.

Housing installation in a new or an existing system is made easy by the band clamp legs that can be adjusted up to 12" in floor–to–outlet height.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty