Would you like to log in again?
3M™ Water Softener System 3MWTS150
Supplier: 3M Healthcare
3M™
7100085395
76028-918EA 5162.28 USD InStock
76028-918
3M™ Water Softener System 3MWTS150
Water Purification Systems Deionizers
3M™ Water Softener System 3MWTS150 treats all incoming water to minimize the effects of hard water typically found in well water. Suitable for water softener system for brighter clothing and cleaner dishes – as well as less detergent, soap and shampoo consumption. Reduced hardness minerals throughout plumbing also decreases scale build-up in plumbing fixtures and water.
  • Meets California Salt Efficiency standards for regulatory compliance
  • Extremely easy-to-use, efficient and state-of-the-art water softening valve
  • Superior performance brine tank featuring a durable, long-lasting design
  • Patented salt shelf protects the valve from dirt and debris and discourages salt bridging
  • Engineered system design helps provide long-term operation
A User-friendly Way to Reduce Hard Water, The 3M™ Water Softener System 3MWTS150 treats all incoming water, minimizing the effects of hard water typically found in well water applications
This system is appropriate for homes with up to three bathrooms. Water often contains calcium, magnesium and other metals that create what’s commonly known as “hard water.” These metal deposits can build up in faucets, pipes and toilets, causing equipment damage and lower water pressure. If left unaddressed, cleanup can become a major hassle and potentially quite expensive. 3M™ Water Softener System 3MWTS150 is an effective, user-friendly solution to this problem. Additional features and specifications include: 1" inlet/outlet connections, 25 – 125 psi/172 – 862 kPa pressure range, 40 – 100 °F/37.8 °C temperature range, Flow rate up to 11.1 gpm/42 lpm, Fully ported four position bypass valve, Meets California Salt Efficiency standards.
Brighter Clothing and Cleaner Dishes, Our 3M™ Water Softener System 3MWTS150 can remove dissolved minerals, produce brighter clothing and cleaner dishes. Treating water with a water softener system can also lead to less detergent, soap and shampoo consumption. Here’s how it works: this system saves water by metering use and regenerating only when necessary. By reducing hardness minerals throughout the plumbing system, scale build-up in plumbing fixtures and water using appliances is reduced. Additionally, the state-of-the-art valve allows monitoring and control of every function and cycle. Users enjoy the valve’s precision, which saves water and energy due to fewer regeneration cycles.
The included multi-level salt shelf, keeps dirt and debris away from the brine draw area, reducing valve maintenance. The brine tank delivers superior performance by featuring a durable, long-lasting design which provides virtually trouble-free operation. Additionally, 3MWTS150 meets the California Salt Efficiency Standards (4000 grains/lb of salt). Collectively, these features are designed for long-term operation to save users time, money and energy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty