Would you like to log in again?
3M™ High Flow Series Chloramines System for Cold Beverage Applications, Model HF295-CL, 5627501
Supplier: 3M Healthcare
3M™
7100074489
76031-712EA 3540.2 USD InStock
76031-712
3M™ High Flow Series Chloramines System for Cold Beverage Applications, Model HF295-CL, 5627501
Filters Filter Cartridges
3M™ High Flow Series Chloramines System for Cold Beverage Applications, Model HF295 – CL provides consistent quality water by reducing particulate*, chloramines, and chlorine taste and odor.
  • Provides consistent quality water
  • Reduces particulate*, chloramines, and chlorine taste and odor from incoming water
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • Designed for chloramine reduction in cold beverage applications
3M™ High Flow Series Chloramines System for Cold Beverage Applications, Model HF295–CL provides consistent quality water by reducing particulate*, chloramines, and chlorine taste and odor
3M™ systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you're giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point–of–entry to dedicated point–of–use water filtration systems, look to 3M Purification to help you find the right solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty