Would you like to log in again?
3M™ High Flow Series Multi-Equipment Chloramines System for Ice, Coffee & Cold Beverage Applications, Model DP295-CL, 5624204
Supplier: 3M Healthcare
3M™
7000125913
76030-494EA 4220.41 USD InStock
76030-494
3M™ High Flow Series Multi-Equipment Chloramines System for Ice, Coffee & Cold Beverage Applications, Model DP295-CL, 5624204
Filters Filter Cartridges
3M™ High Flow Series Multi – Equipment Chloramines System for Ice, Coffee & Cold Beverage Applications, Model DP295 – CL provides consistent quality water by reducing particulate*, chloramines, chlorine taste and odor, and scale*.
  • Provides consistent quality water
  • Reduces particulate*, chloramines, and chlorine taste and odor from incoming water
  • Built–in scale inhibition can help to prevent the potentially harmful effects of scale build–up on equipment
  • Dual Port system is engineered to provide high flow rates, high capacities, and the benefits of multiple–equipment filtration in a single compact package
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • Designed for chloramine reduction in ice, coffee, and cold beverage applications
3M™ High Flow Series Multi–Equipment Chloramines System for Ice, Coffee & Cold Beverage Applications, Model DP295–CL provides consistent quality water by particulate*, chloramines, chlorine taste and odor, and scale*
3M™ systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you're giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point–of–entry to dedicated point–of–use water filtration systems, look to 3M Purification to help you find the right solution.
*As tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty