Would you like to log in again?
3M™ High Flow Series System for Cold Beverage Applications featuring Valve-in-Head Design, Model BEV135, 5616102
Supplier: 3M Healthcare
3M™
7100050156
76030-433CS 3053.02 USD InStock
76030-433
3M™ High Flow Series System for Cold Beverage Applications featuring Valve-in-Head Design, Model BEV135, 5616102
Filters Filter Cartridges
3M™ High Flow Series System for Cold Beverage Applications featuring Valve-in-Head Design, Model BEV135 provides consistent quality water by reducing particulate* and chlorine taste and odor. The Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs without the need for a filter wrench. For use in single carbonator dispensers.
  • Provides consistent quality water
  • Reduces particulate* and chlorine taste and odor from incoming water
  • Valve-in-head design
  • For use in single carbonator dispensers in cold beverage applications
Excess particulate can cause abrasion and short life on pumps, seals, o-rings, and valves, while excess chlorine can cause unpleasant tasting beverages.
Creative Filtration Technology
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
Recipe Quality Water™
Having cleaner and clearer water for your business is both a pleasure and a reassurance that you’re giving your customers recipe quality water. With a water filtration system from 3M Purification, you can provide filtered water that may help protect your equipment and your profits. From commercial point-of-entry to dedicated point-of-use water filtration systems, look to 3M Purification to help you find the right solution.
*As tested and verified by manufacturer’s laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty