Would you like to log in again?
3M™ Aqua-Pure™ Whole House Standard Sump Replacement Water Filter Drop-in Cartridge AP110
Supplier: 3M Healthcare
Aqua-Pure™
7000029441 5620404
76030-477CS 446.04 USD InStock
76030-477 76030-476
3M™ Aqua-Pure™ Whole House Standard Sump Replacement Water Filter Drop-in Cartridge AP110
Filters Filter Cartridges
3M™ Aqua–Pure™ Whole House Standard Diameter Water Filter Drop–in Replacement Cartridge AP110 for the AP100 and standard and large diameter Stainless Steel Housing Series, is suitable for water filtration. This standard 2.5" Ø by 9.8" length drop–in cartridge is manufacturer tested and verified to reduce sediment down to 5 µm nominal.
  • Reduces sediment
  • Average life: 6 months (incoming water quality may affect cartridge life)
  • Proprietary polypropylene manufacturing process
  • Proprietary polypropylene manufacturing process
  • Grooved surface increases total surface area for longer cartridge life than non–grooved filter cartridges
  • Rigid construction helps to eliminate sediment “unloading" due to variances in water pressure
  • System materials are FDA CFR–21 compliant
The 3M™ Aqua–Pure™ Whole House Standard Diameter Water Filter Drop–in Replacement Cartridge AP110 for the AP100 & standard and large diameter Stainless Steel Housing Series, is a great choice in water filtration
Features include: Average life: 6 months (incoming water quality may affect cartridge life), Flow Rate: up to 8 gpm/30.3 lpm (housing dependent), 2.5 "/6.35 cm by 9.8 "/24.9 cm, 25 – 125 psi/172 – 862 kPa pressure range, 4.4 – 37.8 °C (40 – 100 °F
0) temperature range.
To maximize system performance and efficiency, regularly scheduled maintenance including replacement of the filter cartridges at the required interval, at the rated capacity, or sooner if a reduction in water flow occurs is required. Filter life is dependent on actual use and incoming water quality.
3M branded water filtration products have been addressing water quality issues for over 50 years. 3M products offer a wide range of solutions designed to address many types of water quality concerns. Third–party certifications, such as NSF and WQA, are an advantage provided on many 3M Aqua–Pure water quality improvement solutions. 3M, a household brand name, is a known leader in filtration technology recognized for innovation, trust and quality around the world. Having clean and clear drinking water in your home is both a pleasure and a reassurance that you're giving your family the best. With a water filtration system from 3M Purification, you can provide filtered drinking water for every member of your family. From whole home water filtration to drinking water systems, look to 3M Purification to help you find the right solution.
This standard 2.5" diameter drop–in cartridge is manufacturer tested and verified to reduce sediment down to 5–micron nominal. Its rigid construction from our proprietary polypropylene manufacturing process, helps to eliminate “unloading" of sediment due to variances in water pressure, sometimes referred to as water hammer.
Caution: Tested and verified by manufacturer's laboratory.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty