Would you like to log in again?
3M™ Aqua-Pure™ Stainless Steel Whole House System
Supplier: 3M Healthcare
Aqua-Pure™
7000125754 7010409161 7000125753 7000051154 7000051156
76029-030CS 2508.72 USD InStock
76029-030 76029-032 76029-046 76029-034 76029-036
3M™ Aqua-Pure™ Stainless Steel Whole House System
Filters Filter Cartridges
3M™ Aqua–Pure™ Stainless Steel Whole House Filtration System reduces sediment and other contaminants (cartridge dependent) to help provide cleaner, clearer water from the water used throughout your entire home or facility.
  • Stainless steel helps provide maximum strength and durability with very high operating pressure rating
  • Especially suitable for applications where solenoid valves are used
  • Accepts a wide variety of cartridges
  • Enables positive cartridge sealing and easy cartridge changes
  • For high flow rates with minimal pressure drop
  • Please see specification table below for full specs
The 3M™ Aqua–Pure™ SS48 EPE–316L Whole House Filtration System helps provide long lasting, large capacity filtration in harsh corrosive environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty