Would you like to log in again?
3M™ NanoSHIELD™ SDC Series Hollow Fiber Filter Capsule
Supplier: 3M Healthcare
NanoSHIELD™
7010340336 7000051708 7010382797 7010382786 7000051707
76032-602CS 753.64 USD InStock
76032-602 76032-568 76032-578 76032-590 76032-616
3M™ NanoSHIELD™ SDC Series Hollow Fiber Filter Capsule
Filters Capsule Filters
NanoSHIELD hollow fiber series family of products is offered in several configurations to allow the end user to match the volume of fluid with the appropriate filter in an effort to reduce chemical waste.
  • Small, compact design make SDC capsulesIdeal for applications requiring low hold-up volume
  • High particle rentention from 100 nm down to 5 nm
SDC Capsules for advanced photoresists and ancillary chemicals applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty