Would you like to log in again?
3M™ LifeASSURE™ PSN Series Filter Capsule
Supplier: 3M Healthcare
LifeASSURE™
7010412762 7010337865
76034-210CS 459.95 USD InStock
76034-210 76034-212
3M™ LifeASSURE™ PSN Series Filter Capsule
Filters Capsule Filters
Nylon filter capsules feature a highly retentive LifeASSURE PSN filter element designed for photoresist and ancillary chemical applications at point-of-use (POU). Naturally hydrophilic Nylon 6.6 pleated membrane, featuring 3M’s Advanced Pleat Technology, maximizes the surface area of the filter, for excellent gel and particle reduction.
  • Membrane is enclosed in an all-polypropylene capsule construction, available in 2.5 and 5" lengths
  • Available in three grade designations for 0.2, 0.1, and 0.04 µm ratings
  • Allows the filter to be used with no need to pre-wet with IPA and flush
  • Membrane supports are made from high density polyethylene (HDPE)
3M™ LifeASSURE™ PSN Series filter capsules are manufactured using Advanced Pleat Technology, a staggered and stepped configuration which reduces open space between the outside pleats. This novel technology maximizes capacity by increasing the open area, which allows for greater particle loading at the inside diameter, while the shorter stepped pleats take advantage of existing open space closer to the outside diameter of the capsule. The result is a fully used surface area.
Another benefit our Nylon 6.6 membrane offers is a reduction for potential microbubble formation by not de-wetting in outgassing fluids unlike hydrophobic membranes such as polypropylene, UPE, and PTFE. The membrane supports are made from high density polyethylene (HDPE). All capsule components are made of polypropylene and vent O-rings are available in fluorocarbon and EPR. No adhesives, binders, or surfactants are used in the manufacturing process.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty