Would you like to log in again?
3M™ Betapure™ AUL Series Filter Cartridge
Supplier: 3M Healthcare
Betapure™
7010381675 7100046478 7010381673 7010340516 7100057150
76033-665CS 3559.9 USD InStock
76033-665 76032-095 76032-093 76032-097 76033-683
3M™ Betapure™ AUL Series Filter Cartridge
Filters Filter Cartridges
Betapure™ AUL series filters represent a new generation in Chemical Mechanical Planarization (CMP) slurry classification providing superior reduction of large particle counts for improved yields.
  • Rigid, targeted multi–zone structure
  • Consistent filtration from start to finish
  • Superior classification of slurry particles, reducing LPCs
  • Built to controlled specifications to enable repeatable performance in the planarization process
Betapure™ AUL series filters feature 3M's bi–component fiber technology with targeted multi–zone filtration layers to enable Large Particle Count (LPC) reduction and yield improvement in the most advanced CMP processes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty