Would you like to log in again?
3M™ Betapure™ CMP Series Filter Capsule
Supplier: 3M Healthcare
Betapure™
7100005588 7010314318 7100000274 7100054534 7000001786 7000001860 7000001854
76029-838CS 1379.9 USD InStock
76029-838 76029-860 76029-842 76029-844 76029-846 76029-878 76029-836
3M™ Betapure™ CMP Series Filter Capsule
Filters Capsule Filters
Betapure™ CMP filter capsules are high capacity depth filters optimized for oxide and metal slurries used in chemical mechanical planarization (CMP) applications. Betapure CMP filters are composed of all–polypropylene components and feature a multi–zone graded–porosity design for the optimum level of particle classification.
  • Graded porosity design for superior reduction of hard and soft gel contaminants
  • Reduces large particle counts (LPC) that can cause equipment damage and decreased yields
  • Polypropylene depth filter capsules allow for broad chemical and temperature compatibility
  • Manufactured and double–bagged in a clean environment to provide superior downstream cleanliness out of the package
  • Provides a consistent quality of slurry enabling repeatability of the planarization process
  • Fully disposable capsule
Betapure™ CMP series filter capsule offers a graded porosity media that removes particles in sequential size – starting with the larger particles and ending finally with the smallest ones.
It effectively reduces LPC's that can cause equipment damage and potentially reduce yields, while maintaining the polishing characteristics of the slurry. The 100% polypropylene materials offer broad temperature and chemical compatibility, which provides durability throughout demanding process applications.
Recommended Applications: Semiconductor, Oxide/Low–k dielectrics, Shallow Trench Isolation (STI), Inter–level Dielectric (ILD), Polysilicon, Tungsten and copper, Data Storage, Magnetic heads, Nickel and glass substrates, Data Transmission, Optical fiber, Compound Semiconductor, Light Emitting Diodes (LED).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty