Would you like to log in again?
3M™ NanoSHIELD™ LHV Series Hollow Fiber Filter Tube
Supplier: 3M Healthcare
NanoSHIELD™
7100056114
76032-596CS 712.24 USD InStock
76032-596
3M™ NanoSHIELD™ LHV Series Hollow Fiber Filter Tube
Filters Filter Cartridges
NanoSHIELD™ LHV Series hollow fiber filter tubes have been specially designed to provide superior particle retention and low hold-up volume for advanced photoresists and ancillary chemicals.
  • Utilizing Hollow Fiber Technology (HFT), NanoSHIELD hollow fiber filters are able to deliver high flow rates with low hold-up volume, to minimize the waste of experimental photoresist formulations
  • The tubes are offered in ¼ and ⅜" outside diameters thereby allowing common compression fittings to connect directly to the outside of the NanoSHIELD LHV Series hollow fiber tube. In addition, a syringe can also be used to inject fluid directly into the tube
  • NanoSHIELD LHV Series hollow fiber filter tubes contain the same filter media as full size NanoSHIELD hollow fiber series filter cartridges and capsules, ensuring a consistent level of quality throughout laboratory, pilot, and full-scale production testing
  • Hollow Fiber Technology provides more filtration per surface area as compared to pleated filters, and both lower operating and differential pressure across the fi lter to minimize outgassing and micro-bubble formation
  • PP and PE Construction for excellent chemical resistance to photoresists and ancillary chemicals, No IPA pre-wetting and system fl ushing required
  • 0.005 to 0.1 μm Retention Ratings provide superior removal of particles, gels, and micro-bubbles, and reduced micro-bridge and wafer level defects
  • Small, Compact Design reduces hold-up volume and waste of expensive chemicals
LHV tubes for laboratory testing on new formulations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty