Betapure™ AU series filter cartridge in various grades for absolute reduction ratings. The controlled pore size of the filter matrix provides absolute distinction between cartridge grades for accurate and consistent filtration. Filter design provides precise contaminant reduction, consistent effluent quality and superior on–stream service life for a wide–range of applications.
- Rigid structure provides consistent and reproducible contaminant reduction
- Excellent reduction of deformable contaminants for consistent effluent quality
- Gasket design reduces by–pass from poor to damaged seals
- Rigid structure reduces cartridge by–pass and unloading for consistent filtration throughout product lifecycle
- Low pressure drop provides long service life while using smaller filter housings
Our wide range of media grades allows you to precisely tailor performance characteristics and achieve optimal filtration economy by enabling defined reduction cut–off points by particle size
In manufacturing the Betapure™ AU Series Filter we combine stringent material quality assurance standards, exacting in–process controls along with extensive final product testing and verification. The results are products that provide consistent filtration from lot–to–lot, filter–to–filter. The bicomponent fibers of the filter matrix are thermally bonded by exploiting the difference in melt temperatures of the two fiber components. Heating the matrix to the melt temperature of the polyethylene sheath, but below that of the polypropylene core causes the fiber–to–fiber bond at every contact point. And the high degree of fiber–to–fiber bonding provides a rigid structure that eliminates the need for a core support along with any possibility of media migration. Standard Betapure AU Series Filters (polypropylene /polypropylene) are constructed of FDA CFR–21 compliant material.
Applications: Coatings, High Quality Paint, Film Coatings, Resins, and Ink, Food & Beverage, Bottled Water, Pre–Ro, Blend Water, Wash Water, General Industrial, Desalination, Plating, Machine Tool Coolant, Process Water, Electronics, Pre–Ro, Wafer Slurries, Ceramic Slurries, Chemical Mechanical Polishing, Cathode Ray Tube Production, Disc Cleaning, Chemical/Petro–Chemical, Process Water, Pre–Ro, Amine, Fine Chemicals.