Would you like to log in again?
3M™ Betapure™ PK Series Filter Cartridge
Supplier: 3M Healthcare
Betapure™
PK36M100G2VS PK36M100G2WS PK36M300G2WS PK36M300G2NN PK36M200G2WS PK37M200G2RG PK36M100G2NG
76034-087CS 2009.14 USD InStock
76034-087 76034-085 76034-083 76028-603 76034-081 76028-601 76028-605
3M™ Betapure™ PK Series Filter Cartridge
Filters Filter Cartridges
Betapure™ PK series filter cartridges are constructed of cellulose fibers, glass fibers, and a chemically resistant thermosetting resin to produce a durable, rigid filter structure. Standard 2⅝" outside diameter cartridges are grooved to significantly increase the surface area and extend service life. These filter cartridges are compatible in wide range of fluids for oil and gas applications.
  • Filters are absolute rated for reproducible effluent quality throughout the filter's life
  • Grooved surface with true graded–porosity structure increases media surface area
  • No metal or plastic cores for easy shredding
An un–grooved, smooth–surface Betapure PK series filter is available in the same lengths, from ⁹/₃₄" to 40", and absolute ratings from 10 to 60 µm as the grooved version.
It offers a wide range of end–treatments to allow Betapure PK series filter cartridges to easily retrofit installed filter housings. Standard diameter industrial Betapure PK series filter cartridges include polyethylene foam gaskets for positive sealing in double open–end housings. Offers cartridges in the single open–end style that employ integral polypropylene caps and springs, so that separate spring and seal assemblies are not required.
We also use an optimized groove pattern in various Betapure™ PK series filter cartridge models to increase the surface area by over 65% when compared to un–grooved cylindrical cartridges. The grooved surface prevents premature blinding of the outer surface by large particles and allows full utilization of the depth structure. Maximum surface area with a true graded–porosity structure means that Betapure PK series filter cartridges can give you greater service life than standard, un–grooved cartridges. Betapure™ PK series absolute–rated filters provide long on–stream service life to reduce downtime and maintenance and consistent filtration efficiency (Beta–Rated Performance) throughout the entire operation and lower total filtration costs.
Applications: Natural Gas Processing, Amine Sweetening, Glycol Dehydration, Compressor lube oil systems, Fuel Gas, Refining, Amine plant systems, Hydrotreating, Catalytic reforming, Sour water stripping
In addition to the standard style, the Betapure PK Series Filter Cartridge 336 Style is available in a 3" outside diameter in lengths up to 72" and the same absolute ratings as the standard diameter Betapure PK series cartridge. Betapure™ PK series filter cartridges feature a true graded–porosity structure through a manufacturing process that results in a progressively denser center core region creating a graded–porosity structure. Each fiber is locked in place by a thermosetting resin binder to create a rigid depth filter matrix that traps larger particles near the outer surface and smaller particles near the inside diameter. The overall effect can greatly improve cartridge service life by retaining particles and deformable contaminants in decreasing particle size ranges as the contaminant particles progress through the cartridge.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty