Would you like to log in again?
3M™ Micro-Klean™ RB Series Filter Cartridge
Supplier: 3M Healthcare
Micro-Klean™
7100060320
76028-991CS 941.84 USD InStock
76028-991
3M™ Micro-Klean™ RB Series Filter Cartridge
Filters Filter Cartridges
Laboratory testing and field experience has shown Micro-Klean™ RB Series Filter Cartridges hold up to two or more times the contaminant by weight than competitive products with equal retention ratings. Our manufacturing produces a rigid, resin bonded, graded porosity structure that reduces by-pass and avoids the unloading traits of soft, deformable melt blown and string wound filter cartridges.
  • Broad chemical compatibility is ideal for chemically aggressive applications
  • Rigid resin-bonded structure reduces by-pass and avoids unloading with high pressure drops
  • Grooved surface provides 2.3 times more surface area than competitive ungrooved cartridges and provides greater dirt loading capacity
  • Environmental and Energy Advantage with formulation 8 Micro-Klean™ RB Series Filter Cartridges with porosity between 1 and 75 µm contain greater than 20% recycled material by weight
We combine the principles of surface and depth filtration into our Micro-Klean™ RB Series Filter Cartridge line to provide enhanced filter service life, particle removal efficiency and optimum flow characteristics
Standard series filter cartridges are available in this line in multiple lengths with or without various end treatments to fit most major manufacturer's cartridge housings.
Consistent filtration throughout the service life of a depth-style filter depends on how well the filter's structure tolerates fluctuations in operating conditions-including contaminant loading and differential pressure. Flexible structures, such as meltblown and string-wound filters, tend to compress and change porosity with increased pressure, while rigid structures like ours do not. Media compression can result in short filter life because the pores collapse and ultimately close. Media compression can also cause the filter to release already held particles.
Results
The Micro-Klean RB Series Filter Cartridge's extended service life results in fewer filter change-outs while its enhanced flow characteristics can typically reduce the number of filters required to achieve a given flow rate. These combined features can significantly reduce total filtration costs.
Applications include Paints, Inks, Emulsions, Adhesives, Resins, Organic Solvents, Coolants, Lube Oils, Various Chemicals, Pesticides, Fertilizers, Process Water and General Manufacture Processes.
Micro-Klean™ RB Series Filter Cartridges are not designed for and should not be used in food and beverage, and pharmaceutical applications.
Manufacturing process, rigid, resin bonded construction and graded porosity design of our Micro-Klean RB Series Filter Cartridges provides consistency of performance, long filter life and lower overall filtration costs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty