Would you like to log in again?
3M™ 100 Series Bag Filter
Supplier: 3M Healthcare
3M™
7000029513 7000125825 7100006219 7100085491 7000051637 125A 7010339883 7100007394 7000125852 7000125850 7000029498 7010380788 7000051209 7000051249 7010314163 7000051210 7000001843 7000051248
76031-360CS 532.24 USD InStock
76031-360 76031-370 76029-137 76237-842 76031-348 76031-358 76031-328 76031-338 76029-135 76031-344 76031-366 76031-354 76031-332 76031-368 76031-350 76031-363 76031-330 76031-352
3M™ 100 Series Bag Filter
Filters Liquid Process Filters
3M™ 100 Series High Performance Liquid Filter Bags using polypropylene microfibers that allow very fine particle capture at high efficiencies. With over 90% particle removal efficiency at their suggested application rating, our Series 100 filter bags offer an excellent balance of high efficiency with very low initial pressure drop and an effective alternative to filter cartridges.
  • High contaminant holding capacity
  • Can adsorb unwanted trace oils that frequently occur in processed fluids
  • Manufacturing processes do not include the use of silicone or adhesives
Proper sealing of the bag in the filter housing is necessary for prevention of fluid bypass and subsequent contamination of the downstream fluid. 3M 100 Series Liquid Filter Bags are offered in both polypropylene and metal ring sealing collar versions that will properly seal onto most filter bag manufacturer's housings.
Supplied with plastic collar model in which the filter bag media is welded to a specially – designed plastic collar for proper sealing of the 3M™ 100 Series Liquid Filter Bags in most conventional bag housings. The plastic collar incorporates an ergonomically designed handle, which allows easy removal of the filter bag from the filter housing, and for the insertion of bag accessories (displacement balloons, magnets, and more). For ease of identification and traceability, the handle is engraved with micron grade and manufacturing lot information. Ring collar versions use a stainless–steel ring for sealing most versions of the 3M™ 100 Series Liquid Filter Bags into the housing. These versions are appropriate for use in most filter bag housings that will not accept the plastic collar version.
Applications: Acids and bases, machine coolants, amines, make–up water, carbon beds, organic solvents, completion fluids, photo chemicals, deep wells, plating solutions, desalination, RO membranes, DI resins, storm water, glycol, waste water, groundwater clean–up, water flood.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty