Would you like to log in again?
3M™ ZWB Series Housing
Supplier: 3M Healthcare
3M™
7000125841 7000051150 7100066573 7100111016
76029-162CS 17452.46 USD InStock
76029-162 76029-164 76029-166 76029-160
3M™ ZWB Series Housing
Filters Filter Cartridges
3M™ ZWB Series Housing is a multi-cartridge housing designed to accommodate three, five, eight or twelve Single Open End filter cartridges up to 40" long. Three-high housings are the most common with other configurations available upon request. The design uses swing bolt closures to secure the housing dome to the base for operation up to 150 psi (10.3 bar) at 212 °F
  • User friendly vertical design with sanitary connections reduces floor space requirements and reduces disassembly time for cleaning
  • Accepts 3M Purification Inc. sanitary style 226 double O-ring filters, which lock into place assuring secure installation
  • Swing bolt closures offer convenient secure sealing with the housings rated to 150 psi
  • Housings accommodate from three (3) to twelve (12) 10", 20", 30" or 40" cartridges, suitable for a broad range of flow rates
  • Optional sanitary vent valve available provides easy venting
  • Clean-in-place/steam-in-place design permits easy cleaning
Polished sanitary 316L stainless steel construction reduces bacterial/particle adhesion, eases cleaning, and increases resistance to corrosion.
Certifications: 3M™ ZWB housings are designed to meet the requirements of the pharmaceutical, bioprocessing, fine chemical, food and beverage and electronics industries.
Ordering information: 3M ZWB series standard housing models include 2" sanitary Inlet / Outlet (1¹/₂" on 3ZWB), Silicone O-ring, a 1¹/₂" sanitary gauge port and a ¹/₂" sanitary bleed valve connection. An optional ¹/₂" sanitary barb vent/drain diaphragm valve is available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty