Would you like to log in again?
3M™ Original CUNO 8000 Series Replacement Cartridge,Model CFS517LS, 5560012
Supplier: 3M Healthcare
CUNO™
5560012
76030-065CS 1424.75 USD InStock
76030-065
3M™ Original CUNO 8000 Series Replacement Cartridge,Model CFS517LS, 5560012
Filters Filter Cartridges
3M™ Water Filtration Products line of 8000 Series replacement cartridges combine the benefits of filtered water with a design that makes filter replacement quick, clean and easy. Our Sanitary Quick Change (SQC) design allows the operator to never have to touch the media, replacing the entire cartridge with a simple turn. No tools or activation are required.
  • Provides better tasting water for a more consistent product by reducing chlorine taste and odor (excludes CFS8110, CFS8110-S, SWC900 and SWC1350)
  • Improves water clarity by reducing sediment for a variety of commercial applications (excludes CFS8000-S, SWC900, SWC900-C, SWC1350 and SWC1350-C)
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change-outs with a ¼" turn
  • Polyphosphates reduce scale build-up and equipment down time, models CFS8110-S, CFS8112-S, CFS8812X-S, CFS8720-S, CFS517, CFS8000-S, CFS8112EL-S, CFS8112ELX-S and CFS8720EL-S
  • Ion exchange resin reduces hardness without the need for a drain, electrical connection or salt regeneration, models SWC900, SWC900-C, SWC1350 and SWC1350-C
Filter Changes Made Fast and Easy
3M™ Water Filtration Products 8000 Series replacement cartridges make filter change-outs quick and easy. Unlike standard drop-in style filtration cartridges, 8000 series cartridges are manufactured using our Sanitary Quick Change, or SQC, design. The filters’ media is completely encapsulated within the cartridge–the operator never touches either the old or the new replacement filter media, greatly reducing the possibility of contamination. The cartridge installs quickly with as little as a quarter turn, so there is no need for tools.
Foodservice establishment operators can easily change the replacement cartridges themselves, so they can concentrate on other responsibilities–saving them the expense of contacting a service provider. Filters do not require activation and installers do not need to open the filter housing, eliminating the possibility of contamination.
3M™ Water Filtration Products line of 8000 Series replacement cartridges are available for a range of commercial foodservice applications. They are ideal for water filtration for hot coffee and tea, cold beverages and ice machines, and are fully compatible with existing IceAssure™, CFS6000 and CFS7000 Series systems.
NSF Certified Cartridges*
FDA CFR-21 Compliant Material**
Filter cartridges are o-ring seal type
Cartridges are sanitary in design, requiring no contact with the filter media during cartridge change-out.
*3M™ Water Filtration Products replacement cartridge models CFS8112, CFS8112-S, CFS8112EL, CFS8112EL-S, CFS8812X, CFS8812X-S, CFS8812ELX, CFS8812ELX-S, CFS8720, CFS8720-S, CFS517, CFS517LS and CFS8112EL-CL are NSF certified. Visit www.nsf.org for specific claims associated with each product.
**Surfaces that are in contact with water are FDA CFR-21 Compliant.
About Carbon Filtration
Media for 8000 Series cartridges include granular activated carbon (GAC) and graded porosity carbon. One of the most powerful absorbents available, carbon is highly effective due to the vast surface area contained within a small footprint. One pound of carbon contains a surface area of approximately 100 to 125 acres and can absorb an extremely wide range of chemicals. As effective as carbon is, activated carbon is even more effective. Activated carbon has been electro-positively charged to create adsorption. Adsorption is the adhesion of atoms, ions, or molecules from a gas, liquid, or dissolved solid to a surface. This process differs from absorption, where the fluid (absorbate) permeates or is dissolved and held within the filter media. When adsorption is in play, the water, or other fluid to be filtered, passes over the positively charged carbon surface. The negatively charged ions of the contaminants are attracted and held to the surface of the activated carbon particles and effectively removed. Micron ratings for the 8000 Series range from 5 down to 0.5, more than adequate to remove chlorine odor and taste, cysts, sediment and most common bacteria from unfiltered water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty