Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human;Bovine Apelin-16

Supplier: AnaSpec
AS-63699
102971-774EA 185.62 USD InStock
102971-774
Human;Bovine Apelin-16
Proteins and Peptides
It is one of the active peptide fragments resulting from maturation of preproapelin. Apelin family of peptides and receptor has been potentially indicated as target therapeutics of eating disorder and obesity.
Sequence:FRRQRPRLSHKGPMPF
MW:2010.4 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty