Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human;Pig;Rat Viasoactive intestinal peptide (1-12)

Supplier: AnaSpec
AS-24217
103003-696EA 118.38 USD InStock
103003-696
Human;Pig;Rat Viasoactive intestinal peptide (1-12)
Proteins and Peptides
VIP (1−12), vasoactive intestinal peptide fragment 1-12 with a mass of 1425.5 da, is mostly used as a standard in mass spectrometry
Sequence:HSDAVFTDNYTR
MW:1425.5 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty