Would you like to log in again?
Tweezer Repair Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72892-10
103305-758EA 268.34 USD InStock
103305-758
Tweezer Repair Kit, Electron Microscopy Sciences
Forceps
This repair kit is used to straighten, resharpen, and repair tweezers.
Delivery information: This kits includes 1 sharpening stone, 10 O-rings, instrument oil (50 mL) and chain nose pliers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty