Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bovine;Human;Pig Glucagon (1-29)

Supplier: AnaSpec
AS-22456 AS-22457
103003-410EA 248.46 USD InStock
103003-410 103003-408
Bovine;Human;Pig Glucagon (1-29)
Proteins and Peptides
Glucagon is a peptide hormone secreted from the pancreatic Islet of Langerhans alpha-cells, in response to low circulating blood glucose levels in order to restore normal glucose levels. It acts on hepatic enzymes that regulate glucose production and glycogen synthesis. Excessive amounts of circulating glucagon levels is implicated in the metabolic dysregulation of type 2 diabetes, since such conditions result in hyperglycemia.
Sequence: HSQGTFTSDYSKYLDSRRAQDFVQWLMNT
MW: 3482.8 Da
% Peak area by HPLC: 95
Storage condition: -20° C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty