Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Weiler® Extra Heavy-Duty Knot Wire Cup Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-12686
300042-032EA 190.78 USD InStock
300042-032
Weiler® Extra Heavy-Duty Knot Wire Cup Brush, ORS Nasco
Brushes
These brushes are designed for use with right-angled grinders to remove rust, paint, oxides, and scale.
  • Banded
  • Controlled flaring provides high impact action and long life
  • Wire band can be removed once tips are worn down
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty