Would you like to log in again?
Weiler® Stem-Mounted Crimped Wire Cup Brush, ORS Nasco
Supplier: ORS Nasco
804-14301 804-14317 804-14302 804-14306 804-14300 804-14304
300010-177EA 22.04 USD InStock
300010-177 300010-178 300010-179 300046-930 300085-304 300046-932
Weiler® Stem-Mounted Crimped Wire Cup Brush, ORS Nasco
Brushes
These crimped brushes are designed for use with power drills, die grinders, and right-angle air die grinders to remove rust, paint, oxides, and scale.
  • For light to medium duty, general-purpose applications
  • Effective for corners and hard-to-reach areas
  • Cleans rust, paint, and polishes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty